Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTACCCCTTCGGGGGCAGCTACTA[C/T]GGCTGCCGCCTGTCGCACAACGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142976 | ||||||||||||||||||||
Literature Links: |
HOXC13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HOXC13 - homeobox C13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017410.2 | 505 | Silent Mutation | TAC,TAT | Y,Y 130 | NP_059106.2 |
HOXC13-AS - HOXC13 antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |