Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGCGGTGGTGCTAGAATTTGTTA[T/C]ATTTTCCATGAGACTTTTGGGCGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603850 MIM: 610957 | ||||||||||||||||||||
Literature Links: |
DNM1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNM1L - dynamin 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278463.1 | 1268 | Silent Mutation | TAC,TAT | Y,Y 368 | NP_001265392.1 | |
NM_001278464.1 | 1268 | Silent Mutation | TAC,TAT | Y,Y 381 | NP_001265393.1 | |
NM_001278465.1 | 1268 | Silent Mutation | TAC,TAT | Y,Y 381 | NP_001265394.1 | |
NM_001278466.1 | 1268 | Silent Mutation | TAC,TAT | Y,Y 165 | NP_001265395.1 | |
NM_005690.4 | 1268 | Silent Mutation | TAC,TAT | Y,Y 368 | NP_005681.2 | |
NM_012062.4 | 1268 | Silent Mutation | TAC,TAT | Y,Y 368 | NP_036192.2 | |
NM_012063.3 | 1268 | Silent Mutation | TAC,TAT | Y,Y 368 | NP_036193.2 | |
XM_005253282.4 | 1268 | Silent Mutation | TAC,TAT | Y,Y 381 | XP_005253339.1 | |
XM_011520543.2 | 1268 | Silent Mutation | TAC,TAT | Y,Y 381 | XP_011518845.1 | |
XM_011520544.2 | 1268 | Silent Mutation | TAC,TAT | Y,Y 149 | XP_011518846.1 | |
XM_017018663.1 | 1268 | Silent Mutation | TAC,TAT | Y,Y 149 | XP_016874152.1 | |
XM_017018664.1 | 1268 | Silent Mutation | TAC,TAT | Y,Y 149 | XP_016874153.1 | |
XM_017018665.1 | 1268 | Silent Mutation | TAC,TAT | Y,Y 149 | XP_016874154.1 |
YARS2 - tyrosyl-tRNA synthetase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040436.2 | 1268 | Intron | NP_001035526.1 |