Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTCATTGGGTATCTCATGTCAT[T/C]GGTGGGTCTTGGAAAACAAAACAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616146 | ||||||||||||||||||||
Literature Links: |
TGDS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TGDS - TDP-glucose 4,6-dehydratase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304430.1 | 1107 | Missense Mutation | AAT,GAT | N,D 266 | NP_001291359.1 | |
NM_014305.3 | 1107 | Missense Mutation | AAT,GAT | N,D 298 | NP_055120.1 | |
XM_011521065.2 | 1107 | Missense Mutation | AAT,GAT | N,D 266 | XP_011519367.1 | |
XM_011521066.2 | 1107 | Missense Mutation | AAT,GAT | N,D 266 | XP_011519368.1 |