Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCGAGGACCAAGCCCACCCTGGA[C/T]GAGACACCAGAGAACCCACAGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612078 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF469 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF469 - zinc finger protein 469 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127464.2 | 595 | Silent Mutation | GAC,GAT | D,D 134 | NP_001120936.2 | |
XM_017023784.1 | 595 | Silent Mutation | GAC,GAT | D,D 134 | XP_016879273.1 | |
XM_017023785.1 | 595 | Silent Mutation | GAC,GAT | D,D 134 | XP_016879274.1 |