Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCAGCAAATGAGAGTCTGTACCC[G/A]ACCATTTCACAAAAGACCAATGTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603836 MIM: 610355 | ||||||||||||||||||||
Literature Links: |
NDUFAB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NDUFAB1 - NADH:ubiquinone oxidoreductase subunit AB1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PALB2 - partner and localizer of BRCA2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024675.3 | 4227 | Missense Mutation | TCG,TTG | S,L 1165 | NP_078951.2 | |
XM_011545946.2 | 4227 | Missense Mutation | TCG,TTG | S,L 1167 | XP_011544248.1 | |
XM_011545947.2 | 4227 | UTR 3 | XP_011544249.1 | |||
XM_011545948.2 | 4227 | Missense Mutation | TCG,TTG | S,L 870 | XP_011544250.1 | |
XM_017023671.1 | 4227 | Missense Mutation | TCG,TTG | S,L 1088 | XP_016879160.1 | |
XM_017023672.1 | 4227 | Missense Mutation | TCG,TTG | S,L 1086 | XP_016879161.1 | |
XM_017023673.1 | 4227 | UTR 3 | XP_016879162.1 |