Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGGGGCTGCTCCGCGCGCCTGCT[C/T]CCGGGGCTGGCAGGAGGTCCGGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613814 MIM: 614535 | ||||||||||||||||||||
Literature Links: |
TTC19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TTC19 - tetratricopeptide repeat domain 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271420.1 | 556 | UTR 5 | NP_001258349.1 | |||
NM_017775.3 | 556 | Silent Mutation | CTC,CTT | L,L 29 | NP_060245.3 | |
XM_017024801.1 | 556 | Silent Mutation | CTC,CTT | L,L 150 | XP_016880290.1 | |
XM_017024802.1 | 556 | Silent Mutation | CTC,CTT | L,L 150 | XP_016880291.1 |
ZSWIM7 - zinc finger SWIM-type containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042697.1 | 556 | Intron | NP_001036162.1 | |||
NM_001042698.1 | 556 | Intron | NP_001036163.1 |