Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGTCTTCCCTTCAAGGAAGCCA[C/T]GTTTCTAAGAGGGCTCTGGCATATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603494 | ||||||||||||||||||||
Literature Links: |
URI1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
URI1 - URI1, prefoldin like chaperone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252641.1 | 75 | Silent Mutation | CAC,CAT | H,H 11 | NP_001239570.1 | |
NM_003796.3 | 75 | Intron | NP_003787.2 | |||
XM_005259362.2 | 75 | Silent Mutation | CAC,CAT | H,H 11 | XP_005259419.1 | |
XM_005259363.4 | 75 | Intron | XP_005259420.1 | |||
XM_011527435.1 | 75 | Intron | XP_011525737.1 |