Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTCATTCCCCACGATCACCTCA[T/C]TCACTCGTTTAGCTACAAAAGAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606273 MIM: 603673 | ||||||||||||||||||||
Literature Links: |
EIF2B3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EIF2B3 - eukaryotic translation initiation factor 2B subunit gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166588.2 | 1558 | Intron | NP_001160060.1 | |||
NM_001261418.1 | 1558 | UTR 3 | NP_001248347.1 | |||
NM_020365.4 | 1558 | Missense Mutation | AAT,AGT | N,S 440 | NP_065098.1 | |
XM_017002745.1 | 1558 | Intron | XP_016858234.1 | |||
XM_017002746.1 | 1558 | Intron | XP_016858235.1 | |||
XM_017002747.1 | 1558 | Missense Mutation | AAT,AGT | N,S 311 | XP_016858236.1 |
PTCH2 - patched 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |