Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGCGGGCGCGAGCTACGGGTCTG[C/G]AGCGGACCGGGACCGGGACCCGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603051 | ||||||||||||||||||||
Literature Links: |
AGPS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AGPS - alkylglycerone phosphate synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003659.3 | 112 | Missense Mutation | GCA,GGA | A,G 22 | NP_003650.1 | |
XM_011512041.2 | 112 | Intron | XP_011510343.1 | |||
XM_011512042.1 | 112 | Intron | XP_011510344.1 | |||
XM_011512043.2 | 112 | Intron | XP_011510345.1 |
LOC100130691 - uncharacterized LOC100130691 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |