Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGATTGCTCCCCCGCGGCCGCAG[A/G]TCTCCCGTTTGCGCCGCGTTCAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606201 | ||||||||||||||||||||
Literature Links: |
WFS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WFS1 - wolframin ER transmembrane glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145853.1 | 65 | UTR 5 | NP_001139325.1 | |||
NM_006005.3 | 65 | UTR 5 | NP_005996.2 | |||
XM_017008586.1 | 65 | Intron | XP_016864075.1 |