Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTTATGTCCAGATCTTCTACTG[G/A]CACAAGTTCGTGAGTCCCGTGGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603968 MIM: 607845 | ||||||||||||||||||||
Literature Links: |
POLH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLH - DNA polymerase eta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291969.1 | 389 | Nonsense Mutation | TGA,TGG | *,W 26 | NP_001278898.1 | |
NM_001291970.1 | 389 | Missense Mutation | ACA,GCA | T,A 78 | NP_001278899.1 | |
NM_006502.2 | 389 | Missense Mutation | ACA,GCA | T,A 78 | NP_006493.1 | |
XM_005249186.3 | 389 | Missense Mutation | ACA,GCA | T,A 16 | XP_005249243.1 | |
XM_011514698.2 | 389 | Nonsense Mutation | TGA,TGG | *,W 26 | XP_011513000.1 |
XPO5 - exportin 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |