Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGGTGTCGCGGCCGGTCTACAG[C/A]GAGCTCGCTTTCCAGCAACAGCACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605646 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC26A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC26A4 - solute carrier family 26 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000441.1 | 87 | Missense Mutation | AGA,AGC | R,S 28 | NP_000432.1 | |
XM_005250425.2 | 87 | Missense Mutation | AGA,AGC | R,S 28 | XP_005250482.1 | |
XM_006716025.3 | 87 | Missense Mutation | AGA,AGC | R,S 28 | XP_006716088.1 | |
XM_017012318.1 | 87 | Missense Mutation | AGA,AGC | R,S 28 | XP_016867807.1 |
SLC26A4-AS1 - SLC26A4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |