Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCTTAACACTGGAGGCT[GTGAAAACAAC/-]GTCTTTCCCAGGAACTCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614518 MIM: 602136 | ||||||||||||||||||||
Literature Links: |
GATAD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GATAD1 - GATA zinc finger domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021167.4 | 3636 | Intron | NP_066990.3 |
PEX1 - peroxisomal biogenesis factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000466.2 | 3636 | Silent Mutation | NP_000457.1 | |||
NM_001282677.1 | 3636 | Silent Mutation | NP_001269606.1 | |||
NM_001282678.1 | 3636 | Silent Mutation | NP_001269607.1 | |||
XM_017012319.1 | 3636 | Silent Mutation | XP_016867808.1 |