Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601364 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH13 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CDH13 - cadherin 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001220488.1 | Intron | NP_001207417.1 | ||||
NM_001220489.1 | Intron | NP_001207418.1 | ||||
NM_001220490.1 | Intron | NP_001207419.1 | ||||
NM_001220491.1 | Intron | NP_001207420.1 | ||||
NM_001220492.1 | Intron | NP_001207421.1 | ||||
NM_001257.4 | Intron | NP_001248.1 | ||||
XM_011522804.2 | Intron | XP_011521106.1 | ||||
XM_017022848.1 | Intron | XP_016878337.1 | ||||
XM_017022849.1 | Intron | XP_016878338.1 |
Set Membership: |
HapMap Validated |