Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTAAAGCAAATGTCCTATTTCCAT[C/T]ATCCACCCTAAAAATCTGATAATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604619 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LGI1 - leucine rich glioma inactivated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308275.1 | Intron | NP_001295204.1 | ||||
NM_001308276.1 | Intron | NP_001295205.1 | ||||
NM_005097.3 | Intron | NP_005088.1 | ||||
XM_017016911.1 | Intron | XP_016872400.1 | ||||
XM_017016912.1 | Intron | XP_016872401.1 |
LOC101927032 - uncharacterized LOC101927032 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |