Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608866 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRA10AC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FRA10AC1 - fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145246.4 | Intron | NP_660289.2 | ||||
XM_005269515.4 | Intron | XP_005269572.1 | ||||
XM_005269516.4 | Intron | XP_005269573.1 | ||||
XM_011539259.2 | Intron | XP_011537561.1 | ||||
XM_011539260.2 | Intron | XP_011537562.1 | ||||
XM_017015656.1 | Intron | XP_016871145.1 | ||||
XM_017015657.1 | Intron | XP_016871146.1 | ||||
XM_017015658.1 | Intron | XP_016871147.1 | ||||
XM_017015659.1 | Intron | XP_016871148.1 | ||||
XM_017015660.1 | Intron | XP_016871149.1 | ||||
XM_017015661.1 | Intron | XP_016871150.1 | ||||
XM_017015662.1 | Intron | XP_016871151.1 |
Set Membership: |
Validated |