Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGATTGTATTTAGTTGTCATTTA[C/T]TCTTAGTTACAGGTTGTCTGTCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611332 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNAJB6 - DnaJ heat shock protein family (Hsp40) member B6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005494.2 | Intron | NP_005485.1 | ||||
NM_058246.3 | Intron | NP_490647.1 | ||||
XM_005249515.2 | Intron | XP_005249572.1 | ||||
XM_005249516.2 | Intron | XP_005249573.1 | ||||
XM_006715823.1 | Intron | XP_006715886.1 | ||||
XM_011515704.1 | Intron | XP_011514006.1 | ||||
XM_017011633.1 | Intron | XP_016867122.1 |