Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACTCTAAATGAAGAAAAGTCCA[A/G]TGAAGCAAACACGTGACATAAAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605968 MIM: 605972 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RFPL1 - ret finger protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021026.2 | Intron | NP_066306.2 | ||||
XM_011530298.2 | Intron | XP_011528600.1 | ||||
XM_011530299.2 | Intron | XP_011528601.1 | ||||
XM_017028900.1 | Intron | XP_016884389.1 | ||||
XM_017028901.1 | Intron | XP_016884390.1 | ||||
XM_017028902.1 | Intron | XP_016884391.1 |
RFPL1S - RFPL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |