Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATCACTCCTTAAAAATCAATAA[A/G]TGGTGTGAGCATTATTTCACTTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEATR5B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HEATR5B - HEAT repeat containing 5B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019024.2 | 6936 | UTR 3 | NP_061897.1 | |||
XM_006712034.2 | 6936 | UTR 3 | XP_006712097.1 | |||
XM_006712035.3 | 6936 | UTR 3 | XP_006712098.1 | |||
XM_011532934.2 | 6936 | UTR 3 | XP_011531236.1 | |||
XM_011532935.2 | 6936 | UTR 3 | XP_011531237.1 | |||
XM_017004378.1 | 6936 | UTR 3 | XP_016859867.1 | |||
XM_017004379.1 | 6936 | UTR 3 | XP_016859868.1 | |||
XM_017004380.1 | 6936 | UTR 3 | XP_016859869.1 |