Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612471 MIM: 176761 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGXT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGXT2 - alanine--glyoxylate aminotransferase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRLR - prolactin receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000949.6 | 9046 | UTR 3 | NP_000940.1 | |||
NM_001204314.2 | 9046 | UTR 3 | NP_001191243.1 | |||
NM_001204315.1 | 9046 | Intron | NP_001191244.1 | |||
NM_001204316.1 | 9046 | Intron | NP_001191245.1 | |||
NM_001204317.1 | 9046 | Intron | NP_001191246.1 | |||
NM_001204318.1 | 9046 | Intron | NP_001191247.1 | |||
XM_006714484.2 | 9046 | UTR 3 | XP_006714547.1 | |||
XM_011514068.2 | 9046 | UTR 3 | XP_011512370.1 | |||
XM_011514069.2 | 9046 | UTR 3 | XP_011512371.1 | |||
XM_017009645.1 | 9046 | UTR 3 | XP_016865134.1 | |||
XM_017009646.1 | 9046 | Intron | XP_016865135.1 |