Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606420 | ||||||||||||||||||||
Literature Links: |
ELMO1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ELMO1 - engulfment and cell motility 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039459.2 | 4763 | UTR 3 | NP_001034548.1 | |||
NM_001206480.2 | 4763 | UTR 3 | NP_001193409.1 | |||
NM_001206482.1 | 4763 | UTR 3 | NP_001193411.1 | |||
NM_014800.10 | 4763 | UTR 3 | NP_055615.8 | |||
NM_130442.3 | 4763 | UTR 3 | NP_569709.1 | |||
XM_005249919.2 | 4763 | UTR 3 | XP_005249976.1 | |||
XM_006715805.1 | 4763 | UTR 3 | XP_006715868.1 | |||
XM_011515654.1 | 4763 | UTR 3 | XP_011513956.1 | |||
XM_017012838.1 | 4763 | UTR 3 | XP_016868327.1 | |||
XM_017012839.1 | 4763 | UTR 3 | XP_016868328.1 |