Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAGAAGGCCCAGATCTGGGACCC[A/G]GCTCATTTGTTTTCTAGCACTAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600281 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNF4A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HNF4A - hepatocyte nuclear factor 4 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000457.4 | Intron | NP_000448.3 | ||||
NM_001030003.2 | Intron | NP_001025174.1 | ||||
NM_001030004.2 | Intron | NP_001025175.1 | ||||
NM_001258355.1 | Intron | NP_001245284.1 | ||||
NM_001287182.1 | Intron | NP_001274111.1 | ||||
NM_001287183.1 | Intron | NP_001274112.1 | ||||
NM_001287184.1 | Intron | NP_001274113.1 | ||||
NM_175914.4 | Intron | NP_787110.2 | ||||
NM_178849.2 | Intron | NP_849180.1 | ||||
NM_178850.2 | Intron | NP_849181.1 | ||||
XM_005260407.3 | Intron | XP_005260464.1 |