Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610660 MIM: 609771 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLYR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GLYR1 - glyoxylate reductase 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308096.1 | Intron | NP_001295025.1 | ||||
NM_001324096.1 | Intron | NP_001311025.1 | ||||
NM_001324097.1 | Intron | NP_001311026.1 | ||||
NM_001324098.1 | Intron | NP_001311027.1 | ||||
NM_032569.3 | Intron | NP_115958.2 |
UBN1 - ubinuclein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079514.2 | Intron | NP_001072982.1 | ||||
NM_001288656.1 | Intron | NP_001275585.1 | ||||
XM_005255277.3 | Intron | XP_005255334.1 | ||||
XM_005255278.3 | Intron | XP_005255335.1 | ||||
XM_005255279.3 | Intron | XP_005255336.1 | ||||
XM_011522465.1 | Intron | XP_011520767.1 | ||||
XM_011522466.2 | Intron | XP_011520768.1 | ||||
XM_011522467.1 | Intron | XP_011520769.1 | ||||
XM_017023189.1 | Intron | XP_016878678.1 | ||||
XM_017023190.1 | Intron | XP_016878679.1 | ||||
XM_017023191.1 | Intron | XP_016878680.1 | ||||
XM_017023192.1 | Intron | XP_016878681.1 |
Set Membership: |
HapMap Validated |