Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603343 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTRNR2L3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTRNR2L3 - MT-RNR2-like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAE1 - ribonucleic acid export 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015885.1 | Intron | NP_001015885.1 | ||||
NM_003610.3 | Intron | NP_003601.1 | ||||
XM_005260582.2 | Intron | XP_005260639.2 | ||||
XM_005260583.2 | Intron | XP_005260640.1 | ||||
XM_011529087.2 | Intron | XP_011527389.1 | ||||
XM_011529088.2 | Intron | XP_011527390.1 | ||||
XM_011529089.1 | Intron | XP_011527391.1 | ||||
XM_017028108.1 | Intron | XP_016883597.1 |
Set Membership: |
HapMap |