Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGATTTGGAGTGAAGGTGCGGGA[G/A]GCGGTCGAGGCCCTGAGTCTGGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607662 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105372653 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105372653 - uncharacterized LOC105372653 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA2 - spermatogenesis associated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135773.1 | 1550 | Missense Mutation | CTC,TTC | L,F 443 | NP_001129245.1 | |
NM_006038.3 | 1550 | Missense Mutation | CTC,TTC | L,F 443 | NP_006029.1 | |
XM_006723894.1 | 1550 | Missense Mutation | CTC,TTC | L,F 443 | XP_006723957.1 | |
XM_011529116.1 | 1550 | Missense Mutation | CTC,TTC | L,F 443 | XP_011527418.1 |