Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613395 MIM: 600968 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR138-2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR138-2 - microRNA 138-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC12A3 - solute carrier family 12 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000339.2 | Intron | NP_000330.2 | ||||
NM_001126107.1 | Intron | NP_001119579.1 | ||||
NM_001126108.1 | Intron | NP_001119580.1 | ||||
XM_005256119.2 | Intron | XP_005256176.1 |
Set Membership: |
HapMap |