Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGGTGGCTCCTGGTACCGATGG[A/G]GTCCTGACCTACTCAGGGGTGCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616585 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM192A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RSPRY1 - ring finger and SPRY domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305163.1 | 550 | UTR 5 | NP_001292092.1 | |||
NM_001305164.1 | 550 | Intron | NP_001292093.1 | |||
NM_001305182.1 | 550 | Intron | NP_001292111.1 | |||
NM_133368.2 | 550 | Intron | NP_588609.1 | |||
XM_005256220.1 | 550 | Intron | XP_005256277.1 | |||
XM_011523427.1 | 550 | Intron | XP_011521729.1 | |||
XM_011523428.1 | 550 | Intron | XP_011521730.1 | |||
XM_011523430.1 | 550 | Intron | XP_011521732.1 | |||
XM_017023844.1 | 550 | Intron | XP_016879333.1 |