Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAGAAACTTGAGGTTAAGAGTTA[C/T]TGTGTACGTGAATTTTTGTTGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614351 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUP93 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NUP93 - nucleoporin 93 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242795.1 | Intron | NP_001229724.1 | ||||
NM_001242796.1 | Intron | NP_001229725.1 | ||||
NM_014669.4 | Intron | NP_055484.3 | ||||
XM_005256263.3 | Intron | XP_005256320.1 |
Set Membership: |
Validated |