Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAGACCAAAAGAGCAAGAGAAAT[A/G]GAAATGAAAGCAGTAAGTGAATGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605118 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SOCS6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SOCS6 - suppressor of cytokine signaling 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004232.3 | Intron | NP_004223.2 | ||||
XM_005266783.3 | Intron | XP_005266840.1 | ||||
XM_017026086.1 | Intron | XP_016881575.1 | ||||
XM_017026087.1 | Intron | XP_016881576.1 |