Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTATGGCTGGGGGCACTAGGACT[A/G]ACAATCCAGGCAGTCTTTTCCACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C20orf141 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C20orf141 - chromosome 20 open reading frame 141 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256538.1 | 351 | Silent Mutation | CTA,CTG | L,L 59 | NP_001243467.1 | |
NM_080739.2 | 351 | Silent Mutation | CTA,CTG | L,L 59 | NP_542777.1 |
TMEM239 - transmembrane protein 239 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167670.2 | 351 | Intron | NP_001161142.1 | |||
NM_001318207.1 | 351 | Intron | NP_001305136.1 | |||
XM_011529128.2 | 351 | Intron | XP_011527430.1 | |||
XM_011529129.2 | 351 | Intron | XP_011527431.1 |