Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTACACAATTTGCCTGTCCCTTA[A/C]TGATAAAATATTCTGTATCTATACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604570 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHROOM3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SHROOM3 - shroom family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020859.3 | Intron | NP_065910.3 | ||||
XM_005263163.4 | Intron | XP_005263220.1 | ||||
XM_005263164.4 | Intron | XP_005263221.1 | ||||
XM_005263165.4 | Intron | XP_005263222.1 | ||||
XM_006714282.3 | Intron | XP_006714345.1 | ||||
XM_011532157.2 | Intron | XP_011530459.2 | ||||
XM_011532158.2 | Intron | XP_011530460.1 | ||||
XM_011532159.1 | Intron | XP_011530461.1 | ||||
XM_011532160.1 | Intron | XP_011530462.1 | ||||
XM_011532161.2 | Intron | XP_011530463.1 | ||||
XM_017008488.1 | Intron | XP_016863977.1 | ||||
XM_017008489.1 | Intron | XP_016863978.1 |
Set Membership: |
HapMap |