Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTGCCACATTCCAGGCACTGTTG[A/G]GCACTAGGGATACAGCAGAAAACAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602287 MIM: 612755 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERP29 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ERP29 - endoplasmic reticulum protein 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034025.1 | Intron | NP_001029197.1 | ||||
NM_006817.3 | Intron | NP_006808.1 | ||||
XM_017018720.1 | Intron | XP_016874209.1 |
NAA25 - N(alpha)-acetyltransferase 25, NatB auxiliary subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM116 - transmembrane protein 116 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |