Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTATAGTTGACTAATCACACAT[A/C]TCATTGTTTAACTTGACCTGAAAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605753 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPINK2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SPINK2 - serine peptidase inhibitor, Kazal type 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271718.1 | Intron | NP_001258647.1 | ||||
NM_001271719.1 | Intron | NP_001258648.1 | ||||
NM_001271720.1 | Intron | NP_001258649.1 | ||||
NM_001271721.1 | Intron | NP_001258650.1 | ||||
NM_001271722.1 | Intron | NP_001258651.1 | ||||
NM_021114.3 | Intron | NP_066937.1 | ||||
XM_011534405.1 | Intron | XP_011532707.1 | ||||
XM_011534406.2 | Intron | XP_011532708.1 |
Set Membership: |
HapMap |