Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAATTTGGGGCTGTTTAGGATGTG[A/T]AGAAATTATCTCTTGTTCCTGAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607120 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLCB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PLCB1 - phospholipase C beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015192.3 | Intron | NP_056007.1 | ||||
NM_182734.2 | Intron | NP_877398.1 | ||||
XM_011529199.2 | Intron | XP_011527501.1 | ||||
XM_011529200.2 | Intron | XP_011527502.1 | ||||
XM_011529201.2 | Intron | XP_011527503.1 | ||||
XM_011529203.2 | Intron | XP_011527505.1 | ||||
XM_017027752.1 | Intron | XP_016883241.1 | ||||
XM_017027753.1 | Intron | XP_016883242.1 |
Set Membership: |
HapMap Validated |