Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGTGCTCGGGTTGCACGCCCTA[G/T]CGCGGGGGCGACAGCGGCGGCGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 173490 | ||||||||||||||||||||
Literature Links: |
PDGFRA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDGFRA - platelet derived growth factor receptor alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006206.4 | Intron | NP_006197.1 | ||||
XM_005265743.1 | Intron | XP_005265800.1 | ||||
XM_006714039.3 | Intron | XP_006714102.1 | ||||
XM_006714041.3 | Intron | XP_006714104.1 | ||||
XM_011534385.2 | Intron | XP_011532687.1 | ||||
XM_017008280.1 | Intron | XP_016863769.1 | ||||
XM_017008281.1 | Intron | XP_016863770.1 | ||||
XM_017008282.1 | Intron | XP_016863771.1 |