Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTATAATGGTGGAACTTCAGG[C/T]GATAAGCCCAAAAAAGAATGAGATT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604015 | |||||||||||||||||||||||
Literature Links: |
B4GALT4 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
B4GALT4 - beta-1,4-galactosyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003778.3 | Intron | NP_003769.1 | ||||
NM_212543.1 | Intron | NP_997708.1 | ||||
XM_005247855.1 | Intron | XP_005247912.1 | ||||
XM_006713798.2 | Intron | XP_006713861.1 | ||||
XM_006713799.2 | Intron | XP_006713862.1 | ||||
XM_006713800.1 | Intron | XP_006713863.1 | ||||
XM_006713801.2 | Intron | XP_006713864.1 | ||||
XM_011513260.1 | Intron | XP_011511562.1 | ||||
XM_017007401.1 | Intron | XP_016862890.1 |
B4GALT4-AS1 - B4GALT4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |