Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTAGATTTAACAGAATATATGAG[C/T]CTGTAATTAATTAATGGCTCAGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604913 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNOT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNOT7 - CCR4-NOT transcription complex subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322087.1 | 3754 | Intron | NP_001309016.1 | |||
NM_001322088.1 | 3754 | Intron | NP_001309017.1 | |||
NM_001322089.1 | 3754 | Intron | NP_001309018.1 | |||
NM_001322090.1 | 3754 | Intron | NP_001309019.1 | |||
NM_001322091.1 | 3754 | Intron | NP_001309020.1 | |||
NM_001322092.1 | 3754 | Intron | NP_001309021.1 | |||
NM_001322093.1 | 3754 | Intron | NP_001309022.1 | |||
NM_001322094.1 | 3754 | Intron | NP_001309023.1 | |||
NM_001322095.1 | 3754 | Intron | NP_001309024.1 | |||
NM_001322096.1 | 3754 | Intron | NP_001309025.1 | |||
NM_001322097.1 | 3754 | Intron | NP_001309026.1 | |||
NM_001322098.1 | 3754 | Intron | NP_001309027.1 | |||
NM_001322099.1 | 3754 | Intron | NP_001309028.1 | |||
NM_001322100.1 | 3754 | Intron | NP_001309029.1 | |||
NM_013354.6 | 3754 | Intron | NP_037486.2 | |||
NM_054026.3 | 3754 | Intron | NP_473367.2 | |||
XM_005273481.2 | 3754 | UTR 3 | XP_005273538.1 |
ZDHHC2 - zinc finger DHHC-type containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016353.4 | 3754 | Intron | NP_057437.1 |
Set Membership: |
HapMap |