Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601872 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC7A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC7A2 - solute carrier family 7 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008539.3 | Intron | NP_001008539.3 | ||||
NM_001164771.1 | Intron | NP_001158243.1 | ||||
NM_003046.5 | Intron | NP_003037.4 | ||||
XM_005273610.4 | Intron | XP_005273667.1 | ||||
XM_005273611.4 | Intron | XP_005273668.1 | ||||
XM_005273612.4 | Intron | XP_005273669.1 | ||||
XM_017013746.1 | Intron | XP_016869235.1 | ||||
XM_017013747.1 | Intron | XP_016869236.1 |