Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGAGGGAGCTGAGGGAACTCGGG[C/G]AGGATTCATAAGGGGCCCAGATCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600249 MIM: 610803 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH1L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ALDH1L1 - aldehyde dehydrogenase 1 family member L1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270364.1 | Intron | NP_001257293.1 | ||||
NM_001270365.1 | Intron | NP_001257294.1 | ||||
NM_012190.3 | Intron | NP_036322.2 | ||||
XM_006713481.2 | Intron | XP_006713544.1 | ||||
XM_011512355.1 | Intron | XP_011510657.1 | ||||
XM_017005613.1 | Intron | XP_016861102.1 | ||||
XM_017005614.1 | Intron | XP_016861103.1 |
ALDH1L1-AS1 - ALDH1L1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC41A3 - solute carrier family 41 member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |