Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609739 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ILDR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ILDR1 - immunoglobulin like domain containing receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199799.1 | 1490 | Silent Mutation | CTG,CTT | L,L 515 | NP_001186728.1 | |
NM_001199800.1 | 1490 | Silent Mutation | CTG,CTT | L,L 426 | NP_001186729.1 | |
NM_175924.3 | 1490 | Silent Mutation | CTG,CTT | L,L 471 | NP_787120.1 | |
XM_005247389.4 | 1490 | Silent Mutation | CTG,CTT | L,L 483 | XP_005247446.1 | |
XM_011512738.2 | 1490 | Silent Mutation | CTG,CTT | L,L 515 | XP_011511040.1 | |
XM_011512739.2 | 1490 | Silent Mutation | CTG,CTT | L,L 336 | XP_011511041.1 |
Set Membership: |
HapMap |