Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607659 MIM: 602339 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EAF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EAF2 - ELL associated factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC15A2 - solute carrier family 15 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145998.1 | 35 | UTR 5 | NP_001139470.1 | |||
NM_021082.3 | 35 | UTR 5 | NP_066568.3 | |||
XM_005247722.3 | 35 | Intron | XP_005247779.1 | |||
XM_006713736.3 | 35 | Intron | XP_006713799.1 | |||
XM_017007074.1 | 35 | Intron | XP_016862563.1 |