Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAACAATATATTTACCTTCCACAT[A/G]AGTTTTTATTGATACCAAGATGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611170 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAMD9L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SAMD9L - sterile alpha motif domain containing 9 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303496.1 | 5254 | UTR 3 | NP_001290425.1 | |||
NM_001303497.1 | 5254 | UTR 3 | NP_001290426.1 | |||
NM_001303498.1 | 5254 | UTR 3 | NP_001290427.1 | |||
NM_001303500.1 | 5254 | UTR 3 | NP_001290429.1 | |||
NM_152703.3 | 5254 | UTR 3 | NP_689916.2 | |||
XM_005250193.3 | 5254 | UTR 3 | XP_005250250.1 | |||
XM_006715890.1 | 5254 | UTR 3 | XP_006715953.1 | |||
XM_011515903.2 | 5254 | UTR 3 | XP_011514205.1 | |||
XM_017011822.1 | 5254 | UTR 3 | XP_016867311.1 | |||
XM_017011823.1 | 5254 | UTR 3 | XP_016867312.1 |