Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAGTTCAGTCTGCAGTGATGGC[A/G]GGAACAATCTCAGGAGTCTGGAAGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
PXYLP1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PXYLP1 - 2-phosphoxylose phosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037172.2 | Intron | NP_001032249.1 | ||||
NM_001282728.1 | Intron | NP_001269657.1 | ||||
NM_152282.4 | Intron | NP_689495.1 | ||||
XM_005247903.3 | Intron | XP_005247960.1 | ||||
XM_011513314.2 | Intron | XP_011511616.1 | ||||
XM_017007510.1 | Intron | XP_016862999.1 | ||||
XM_017007511.1 | Intron | XP_016863000.1 |