Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607989 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPOCK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SPOCK3 - sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040159.1 | 1607 | UTR 3 | NP_001035249.1 | |||
NM_001204352.1 | 1607 | UTR 3 | NP_001191281.1 | |||
NM_001204353.1 | 1607 | UTR 3 | NP_001191282.1 | |||
NM_001204354.1 | 1607 | UTR 3 | NP_001191283.1 | |||
NM_001204355.1 | 1607 | UTR 3 | NP_001191284.1 | |||
NM_001204356.1 | 1607 | UTR 3 | NP_001191285.1 | |||
NM_001204358.1 | 1607 | Intron | NP_001191287.1 | |||
NM_001251967.1 | 1607 | UTR 3 | NP_001238896.1 | |||
NM_016950.2 | 1607 | UTR 3 | NP_058646.2 | |||
XM_011532018.1 | 1607 | UTR 3 | XP_011530320.1 | |||
XM_017008257.1 | 1607 | UTR 3 | XP_016863746.1 | |||
XM_017008258.1 | 1607 | UTR 3 | XP_016863747.1 |
Set Membership: |
HapMap Validated |