Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616919 | ||||||||||||||||||||
Literature Links: |
FRMPD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FRMPD1 - FERM and PDZ domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014907.2 | Intron | NP_055722.2 | ||||
XM_011517803.2 | Intron | XP_011516105.1 | ||||
XM_011517804.2 | Intron | XP_011516106.1 | ||||
XM_011517805.2 | Intron | XP_011516107.1 | ||||
XM_011517806.2 | Intron | XP_011516108.1 | ||||
XM_017014480.1 | Intron | XP_016869969.1 | ||||
XM_017014481.1 | Intron | XP_016869970.1 | ||||
XM_017014482.1 | Intron | XP_016869971.1 |
RAB1C - RAB1C, member RAS oncogene family pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |