Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCAGTGGCTGGGGGGTTGAAAA[C/T]CCTTGGCTTTGTTGGCTTAACATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613578 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRSS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRSS3 - protease, serine 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197097.2 | Intron | NP_001184026.2 | ||||
NM_001197098.1 | Intron | NP_001184027.1 | ||||
NM_002771.3 | Intron | NP_002762.2 | ||||
NM_007343.3 | Intron | NP_031369.2 | ||||
XM_011517965.1 | Intron | XP_011516267.1 |
Set Membership: |
HapMap |