Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191760 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UGP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
UGP2 - UDP-glucose pyrophosphorylase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001521.1 | Intron | NP_001001521.1 | ||||
NM_006759.3 | Intron | NP_006750.3 | ||||
XM_005264537.2 | Intron | XP_005264594.1 | ||||
XM_005264538.1 | Intron | XP_005264595.1 | ||||
XM_006712088.1 | Intron | XP_006712151.1 | ||||
XM_011533091.1 | Intron | XP_011531393.1 | ||||
XM_017004855.1 | Intron | XP_016860344.1 | ||||
XM_017004856.1 | Intron | XP_016860345.1 | ||||
XM_017004857.1 | Intron | XP_016860346.1 |