Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTGGCTGGAATAGACTAGTACCA[C/G]CATTTTTGTTGGCAATGCAGTAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBM27 - RNA binding motif protein 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018989.1 | Intron | NP_061862.1 | ||||
XM_005268466.1 | Intron | XP_005268523.1 | ||||
XM_005268467.1 | Intron | XP_005268524.1 | ||||
XM_005268468.1 | Intron | XP_005268525.1 | ||||
XM_006714787.1 | Intron | XP_006714850.1 | ||||
XM_011537661.1 | Intron | XP_011535963.1 | ||||
XM_011537662.1 | Intron | XP_011535964.1 | ||||
XM_017009587.1 | Intron | XP_016865076.1 |