Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120270 MIM: 604745 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL9A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
COL9A3 - collagen type IX alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001853.3 | 2327 | Intron | NP_001844.3 | |||
XM_011528545.1 | 2327 | Intron | XP_011526847.1 | |||
XM_017027666.1 | 2327 | Intron | XP_016883155.1 |
DPH3P1 - diphthamide biosynthesis 3 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCFL5 - transcription factor like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301726.1 | 2327 | Intron | NP_001288655.1 | |||
NM_006602.3 | 2327 | UTR 3 | NP_006593.2 | |||
XM_005260185.3 | 2327 | UTR 3 | XP_005260242.1 | |||
XM_011528497.2 | 2327 | Intron | XP_011526799.1 | |||
XM_011528498.2 | 2327 | Intron | XP_011526800.1 | |||
XM_011528499.1 | 2327 | Intron | XP_011526801.1 |
Set Membership: |
HapMap Validated |