Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAAGACAACTTCCCTGGAATCC[A/G]TTTTTTTCTCCAATCTACTATTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609383 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102724404 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC102724404 - uncharacterized LOC102724404 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NIPAL4 - NIPA like domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099287.1 | Intron | NP_001092757.1 | ||||
NM_001172292.1 | Intron | NP_001165763.1 | ||||
XM_011534552.1 | Intron | XP_011532854.1 |